missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzoic Acid, TRC

Product Code. 30353777
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30353777 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30353777 Supplier Toronto Research Chemicals Supplier No. TRCC36465025G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Chlorobenzoic acid
CAS 74-11-3
Molecular Formula C7 H5 Cl O2
Quantity 25 g
Synonym Benzoic acid, p-chloro- (7CI,8CI), 4-Chlorobenzoic acid, 4-CBA, Chlorobenzene-4-carboxylic acid, Mycosid, NSC 143358, NSC 32738, NSC 8444, p-Carboxychlorobenzene, p-Chlorbenzoic acid, p-Chlorobenzoic acid
InChI Formula InChI=1S/C7H5ClO2/c8-6-3-1-5(2-4-6)7(9)10/h1-4H,(H,9,10)
SMILES OC(=O)c1ccc(Cl)cc1
IUPAC Name 4-chlorobenzoic acid
Molecular Weight (g/mol) 156.57
Formula Weight 155.9978
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.