missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzhydrazide, 98%

Product Code. 11403894
Change view
Click to view available options
Quantity:
5 g
50 g
100 g
Unit Size:
100g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403894 5 g 5g
11304759 50 g 50g
11413894 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11403894 Supplier Thermo Scientific Alfa Aesar Supplier No. A11911.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 536-40-3 | C7H7ClN2O | 170.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 536-40-3
Molecular Formula C7H7ClN2O
Molecular Weight (g/mol) 170.60
MDL Number MFCD00007603
InChI Key PKBGHORNUFQAAW-UHFFFAOYSA-N
Synonym 4-chlorobenzhydrazide, 4-chlorobenzoylhydrazine, 4-chlorobenzoic hydrazide, p-chlorobenzohydrazide, 4-chlorobenzoic acid hydrazide, p-chlorobenzoylhydrazine, p-chlorobenzoic hydrazide, p-chlorobenzoyl hydrazide, 4-chlorobenzoyl hydrazide, p-chlorobenzoic acid, hydrazide
PubChem CID 10816
IUPAC Name 4-chlorobenzohydrazide
SMILES NNC(=O)C1=CC=C(Cl)C=C1

Specifications

Melting Point 162°C to 166°C
Quantity 5 g
Beilstein 511154
Formula Weight 170.6
Percent Purity 98%
Chemical Name or Material 4-Chlorobenzhydrazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.