missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzenesulfonyl Chloride, TRC

Product Code. 30362220
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30362220 10 g 10g
30352923 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30362220 Supplier Toronto Research Chemicals Supplier No. TRCC25349610G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Chlorobenzenesulfonyl Chloride
CAS 98-60-2
Molecular Formula C6H4Cl2O2S
Quantity 10 g
Synonym 4-Chlorobenzene-1-sulfonyl Chloride, 4-Chlorophenylsulfonyl Chloride, NSC 6956, p-Chlorobenzenesulfonyl Chloride, p-Chlorophenylsulfonyl Chloride
InChI Formula InChI=1S/C6H4Cl2O2S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4H
SMILES Clc1ccc(cc1)S(=O)(=O)Cl
IUPAC Name 4-chlorobenzenesulfonyl chloride
Molecular Weight (g/mol) 211.07
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.