missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzenesulfonyl chloride, 97%

Product Code. 10367620 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10367620 100 g Plastic bottle 100g
10407190 5 g Glass bottle 5g
10714661 25 g Glass bottle 25g
10032020 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10367620 Supplier Thermo Scientific Acros Supplier No. 158561000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 98-60-2 | C6H4Cl2O2S | 211.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-60-2
Molecular Formula C6H4Cl2O2S
Molecular Weight (g/mol) 211.06
MDL Number MFCD00007439
InChI Key ZLYBFBAHAQEEQQ-UHFFFAOYSA-N
Synonym 4-chlorobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-chloro, p-chlorophenylsulfonyl chloride, p-chlorobenzenesulfonyl chloride, 4-chlorobenzenesulfonylchloride, benzenesulfonyl chloride, p-chloro, p-chlorbenzensulfochlorid, 4-chlorophenylsulfonyl chloride, 4-chlorobenzenesulphonyl chloride, 4-chloro-benzenesulfonyl chloride
PubChem CID 7398
IUPAC Name 4-chlorobenzenesulfonyl chloride
SMILES ClC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 50°C to 52°C
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Plastic bottle
Linear Formula ClC6H4SO2Cl
Quantity 100 g
Beilstein 11, 55
Solubility Information Solubility in water: insoluble in water
Formula Weight 211.07
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Chlorobenzenesulfonyl chloride, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chlorobenzenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.