missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chlorobenzenesulfonyl chloride, 97%

Product Code. 11493993
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11493993 500 g 500g
11483993 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11493993 Supplier Alfa Aesar™ Supplier No. A10271.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 98-60-2
Molecular Formula C6H4Cl2O2S
Molecular Weight (g/mol) 211.06
MDL Number MFCD00007439
InChI Key ZLYBFBAHAQEEQQ-UHFFFAOYSA-N
Synonym 4-chlorobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-chloro, p-chlorophenylsulfonyl chloride, p-chlorobenzenesulfonyl chloride, 4-chlorobenzenesulfonylchloride, benzenesulfonyl chloride, p-chloro, p-chlorbenzensulfochlorid, 4-chlorophenylsulfonyl chloride, 4-chlorobenzenesulphonyl chloride, 4-chloro-benzenesulfonyl chloride
PubChem CID 7398
IUPAC Name 4-chlorobenzene-1-sulfonyl chloride
SMILES ClC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Melting Point 49°C to 54°C
Boiling Point 141°C (15 mmHg)
Flash Point 107°C (224°F)
Quantity 500 g
UN Number UN3261
Beilstein 511583
Formula Weight 211.07
Percent Purity 97%
Chemical Name or Material 4-Chlorobenzenesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.