missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzenesulfinic acid sodium salt hydrate, 97%

Product Code. 11464294
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464294 25 g 25g
11454294 5 g 5g
11474294 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464294 Supplier Thermo Scientific Alfa Aesar Supplier No. A12583.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14752-66-0 | C6H4ClNaO2S | 198.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14752-66-0
Molecular Formula C6H4ClNaO2S
Molecular Weight (g/mol) 198.60
MDL Number MFCD00035602
InChI Key JFXAUUFCZJYLJF-UHFFFAOYSA-M
Synonym sodium 4-chlorobenzenesulfinate, 4-chlorobenzenesulfinic acid sodium salt, sodium 4-chlorobenzene sulfinate, unii-x2u129fz4n, sodium p-chlorobenzenesulphinate, sodium 4-chlorobenzene-1-sulfinate, 4-chlorobenzene sulfinic acid sodium, 4-chlorobenzenesulfinic acid sodium salt hydrate, p-chlorobenzenesulfinic acid, sodium salt, acmc-1cin1
PubChem CID 23664783
IUPAC Name sodium;4-chlorobenzenesulfinate
SMILES [Na+].[O-]S(=O)C1=CC=C(Cl)C=C1

Specifications

Melting Point >300°C
Quantity 25 g
Beilstein 3575604
Formula Weight 198.60 (Anhydrous)
Percent Purity 97%
Chemical Name or Material 4-Chlorobenzenesulfinic acid sodium salt hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.