missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzeneboronic Acid, TRC

Product Code. 30349198
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30349198 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30349198 Supplier Toronto Research Chemicals Supplier No. TRCC36456510G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Chlorobenzeneboronic Acid
CAS 1679-18-1
Molecular Formula C6H6BClO2
Quantity 10 g
Synonym B-(4-chlorophenyl)boronic Acid, p-Chlorobenzeneboronic Acid, (4-Chlorophenyl)boronic Acid, (p-Chlorophenyl)boronic Acid, 4-Chlorobenzeneboronic Acid, NSC 25408, p-Chlorobenzeneboronic Acid
InChI Formula InChI=1S/C6H6BClO2/c8-6-3-1-5(2-4-6)7(9)10/h1-4,9-10H
SMILES OB(O)c1ccc(Cl)cc1
IUPAC Name (4-chlorophenyl)boronic acid
Molecular Weight (g/mol) 156.37
Formula Weight 156.0149
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.