missing translation for 'onlineSavingsMsg'
Learn More

4-Chloroanisole, 99%, Thermo Scientific Chemicals

Catalog Number 11460193
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
623-12-1
C7H7ClO
142.582
MFCD00000616
YRGAYAGBVIXNAQ-UHFFFAOYSA-N
4-chloroanisole, benzene, 1-chloro-4-methoxy, p-chloroanisole, p-chlorophenyl methyl ether, anisyl chloride, anisole, p-chloro, p-chloromethoxybenzene, 4-chlorophenol methyl ether, 1-chloro-4-methoxy-benzene, unii-0f18bly08q
12167
1-chloro-4-methoxybenzene
COC1=CC=C(C=C1)Cl
This item is not returnable. View return policy
623-12-1
C7H7ClO
142.582
MFCD00000616
YRGAYAGBVIXNAQ-UHFFFAOYSA-N
4-chloroanisole, benzene, 1-chloro-4-methoxy, p-chloroanisole, p-chlorophenyl methyl ether, anisyl chloride, anisole, p-chloro, p-chloromethoxybenzene, 4-chlorophenol methyl ether, 1-chloro-4-methoxy-benzene, unii-0f18bly08q
12167
1-chloro-4-methoxybenzene
COC1=CC=C(C=C1)Cl

4-Chloroanisole was found to react with Cu (II)-smectite forming a blue clay-organic complex. 4-Chloroanisole was chosen as the main test substrate for the optimisation studies as it is electronically deactivated and thus resistant to oxidative addition and consequently very reluctant to enter a catalytic manifold.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloroanisole was found to react with Cu (II)-smectite forming a blue clay-organic complex. 4-Chloroanisole was chosen as the main test substrate for the optimisation studies as it is electronically deactivated and thus resistant to oxidative addition and consequently very reluctant to enter a catalytic manifold.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point -18°C
Density 1.17
Boiling Point 198°C to 200°C
Flash Point 78°C (172°F)
Refractive Index 1.535
Quantity 250 g
Beilstein 1858901
Solubility Information Insoluble in water.
Formula Weight 142.59
Percent Purity 99%
Chemical Name or Material 4-Chloroanisole
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'