missing translation for 'onlineSavingsMsg'
Learn More

4-Chloroanisole, 99%

Product Code. 11440193
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440193 1000 g 1000g
11450193 50 g 50g
11460193 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440193 Supplier Thermo Scientific Alfa Aesar Supplier No. A11377.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 623-12-1 | C7H7ClO | 142.582 g/mol

4-Chloroanisole was found to react with Cu (II)-smectite forming a blue clay-organic complex. 4-Chloroanisole was chosen as the main test substrate for the optimisation studies as it is electronically deactivated and thus resistant to oxidative addition and consequently very reluctant to enter a catalytic manifold.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloroanisole was found to react with Cu (II)-smectite forming a blue clay-organic complex. 4-Chloroanisole was chosen as the main test substrate for the optimisation studies as it is electronically deactivated and thus resistant to oxidative addition and consequently very reluctant to enter a catalytic manifold.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-12-1
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.582
MDL Number MFCD00000616
InChI Key YRGAYAGBVIXNAQ-UHFFFAOYSA-N
Synonym 4-chloroanisole, benzene, 1-chloro-4-methoxy, p-chloroanisole, p-chlorophenyl methyl ether, anisyl chloride, anisole, p-chloro, p-chloromethoxybenzene, 4-chlorophenol methyl ether, 1-chloro-4-methoxy-benzene, unii-0f18bly08q
PubChem CID 12167
IUPAC Name 1-chloro-4-methoxybenzene
SMILES COC1=CC=C(C=C1)Cl

Specifications

Melting Point -18°C
Density 1.17
Boiling Point 198°C to 200°C
Flash Point 78°C (172°F)
Refractive Index 1.535
Quantity 1000 g
Beilstein 1858901
Solubility Information Insoluble in water.
Formula Weight 142.59
Percent Purity 99%
Chemical Name or Material 4-Chloroanisole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.