missing translation for 'onlineSavingsMsg'
Learn More

4-chloroaniline hydrochloride, TRC

Product Code. 30365366
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30365366 50 g 50g
30363574 10 g 10g
30355848 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30365366 Supplier Toronto Research Chemicals Supplier No. TRCC46031550G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20265-96-7
Molecular Formula C6 H6 Cl N . Cl H
Molecular Weight (g/mol) 164.03
Synonym Benzenamine, 4-chloro-, hydrochloride (1:1), Aniline, p-chloro-, hydrochloride (8CI), Benzenamine, 4-chloro-, hydrochloride (9CI), 4-Chloroaniline hydrochloride, 4-Chloroaniline monohydrochloride, 4-Chloroanilinium chloride, 4-Chlorobenzenamine hydrochloride, 4-Chlorophenylammonium chloride, p-Chloroaniline hydrochloride, p-Chloroanilinium chloride, p-Chlorophenylamine hydrochloride
IUPAC Name 4-chloroaniline;hydrochloride
SMILES Cl.Nc1ccc(Cl)cc1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C6H6ClN.ClH/c7-5-1-3-6(8)4-2-5;/h1-4H,8H2;1H
Formula Weight 162.9956
Chemical Name or Material p-Chloroaniline hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.