missing translation for 'onlineSavingsMsg'
Learn More

4-Chloroaniline hydrochloride, 97%, Thermo Scientific™

Product Code. 11333217
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11333217 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11333217 Supplier Thermo Scientific Alfa Aesar Supplier No. L13744.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20265-96-7
Molecular Formula C6H7Cl2N
Molecular Weight (g/mol) 164.029
MDL Number MFCD00042057
InChI Key ISJBQSJDQZLCSF-UHFFFAOYSA-N
Synonym 4-chloroaniline hydrochloride, p-chloroaniline hydrochloride, 4-chlorobenzenamine hydrochloride, 4-chloroanilinium chloride, benzenamine, 4-chloro-, hydrochloride, p-chloroanilinium chloride, unii-po3w01cfsw, p-chloroaniline hcl, 4-chloroaniline hcl, p-chlorophenylamine hydrochloride
PubChem CID 5284363
IUPAC Name 4-chloroaniline;hydrochloride
SMILES C1=CC(=CC=C1N)Cl.Cl

Specifications

Melting Point >230°C (sublimation)
Odor Characteristic
Quantity 25 g
UN Number UN2018
Beilstein 3560964
Formula Weight 164.03
Percent Purity 97%
Chemical Name or Material 4-Chloroaniline hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.