missing translation for 'onlineSavingsMsg'
Learn More

4-Chloroaniline, 98%

Product Code. 10090740 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic Bottle
Unit Size:
100g
1kg
500g
5g
5kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10090740 500 g Glass bottle 500g
10080740 5 g Glass bottle 5g
10677632 100 g Glass bottle 100g
10355930 1 kg Glass bottle 1kg
10267263 5 kg Plastic Bottle 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10090740 Supplier Thermo Scientific Acros Supplier No. 108595000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 106-47-8 | C6H6ClN | 127.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-47-8
Molecular Formula C6H6ClN
Molecular Weight (g/mol) 127.57
MDL Number MFCD00007835
InChI Key QSNSCYSYFYORTR-UHFFFAOYSA-N
Synonym p-chloroaniline, 4-chlorobenzenamine, p-chloraniline, benzenamine, 4-chloro, p-chlorophenylamine, 4-chlorophenylamine, para-chloroaniline, 1-amino-4-chlorobenzene, p-aminochlorobenzene, 4-aminochlorobenzene
PubChem CID 7812
ChEBI CHEBI:20331
IUPAC Name 4-chloroaniline
SMILES C1=CC(=CC=C1N)Cl

Specifications

Melting Point 68.0°C to 72.0°C
CAS Min % 97.5
Color Beige to Brown-Purple
CAS Max % 100.0
Quantity 500 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Solid
Assay Percent Range 97.5% min. (GC)
Linear Formula ClC6H4NH2
Viscosity 1.2 mPa.s (90°C)
Beilstein 12,607
Merck Index 15,2118
Boiling Point 232.0°C
Solubility Information Solubility in water: 3g/L in water (20°C). Other solubilities: freely soluble in alcohol, ether, acetone and, carbon disulfide
Formula Weight 127.57
Percent Purity 98%
Chemical Name or Material 4-Chloroaniline, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.