missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloroacetanilide, TRC

Product Code. 17994025
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17994025 5 g 5g
17910985 1 g 1g
17922005 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17994025 Supplier Toronto Research Chemicals Supplier No. TRCC3633555G
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 539-03-7
Molecular Formula C8 H8 Cl N O
Molecular Weight (g/mol) 169.61
Synonym Acetanilide, 4'-chloro- (8CI), N-(4-Chlorophenyl)acetamide, 4'-Chloroacetanilide, N-(4-Chlorophenyl)ethaneamide, N-(p-Chlorophenyl)acetamide, N-Acetyl-4-chloroaniline, N-Acetyl-p-chloroaniline, NSC 40563, NSC 444, p-Chloroacetanilide
IUPAC Name N-(4-chlorophenyl)acetamide
SMILES CC(=O)Nc1ccc(Cl)cc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C8H8ClNO/c1-6(11)10-8-4-2-7(9)3-5-8/h2-5H,1H3,(H,10,11)
Formula Weight 169.0294
Percent Purity >95%
Chemical Name or Material 4-Chloroacetanilide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.