missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloroacetanilide, 98+%

Product Code. 11433534
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433534 500 g 500g
11324759 25 g 25g
11423534 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11433534 Supplier Thermo Scientific Alfa Aesar Supplier No. A11214.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 539-03-7 | C8H8ClNO | 169.608 g/mol

4'-Chloroacetanilide is used in pharmaceutical preparations, using PGC column. And also used in the synthesis of alprazolam.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Chloroacetanilide is used in pharmaceutical preparations, using PGC column. And also used in the synthesis of alprazolam.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 539-03-7
Molecular Formula C8H8ClNO
Molecular Weight (g/mol) 169.608
MDL Number MFCD00000612
InChI Key GGUOCFNAWIODMF-UHFFFAOYSA-N
Synonym 4'-chloroacetanilide, n-4-chlorophenyl acetamide, 4-chloroacetanilide, p-chloroacetanilide, n-acetyl-p-chloroaniline, acetamide, n-4-chlorophenyl, acetanilide, 4'-chloro, acetic-4-chloroanilide, n-p-chlorophenyl acetamide, n-4-chloro-phenyl-acetamide
PubChem CID 10871
ChEBI CHEBI:116915
IUPAC Name N-(4-chlorophenyl)acetamide
SMILES CC(=O)NC1=CC=C(C=C1)Cl

Specifications

Melting Point 178°C to 180°C
Density 1.385
Boiling Point 333°C
Quantity 500 g
Beilstein 509638
Merck Index 14,2111
Solubility Information Insoluble in water.
Formula Weight 169.61
Percent Purity ≥98%
Chemical Name or Material 4'-Chloroacetanilide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.