missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloroacetanilide, 97%

Product Code. 10742211 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
250g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10742211 50 g Plastic bottle 50g
10732211 5 g Glass Bottle 5g
10188133 250 g Plastic bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10742211 Supplier Thermo Scientific Acros Supplier No. 169950500
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 539-03-7 | C8H8ClNO | 169.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 539-03-7
MDL Number MFCD00000612
InChI Key GGUOCFNAWIODMF-UHFFFAOYSA-N
Synonym 4'-chloroacetanilide, n-4-chlorophenyl acetamide, 4-chloroacetanilide, p-chloroacetanilide, n-acetyl-p-chloroaniline, acetamide, n-4-chlorophenyl, acetanilide, 4'-chloro, acetic-4-chloroanilide, n-p-chlorophenyl acetamide, n-4-chloro-phenyl-acetamide
PubChem CID 10871
ChEBI CHEBI:116915
IUPAC Name N-(4-chlorophenyl)acetamide
SMILES CC(=O)NC1=CC=C(C=C1)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 177°C to 181°C
Color Beige-Gray or White
Boiling Point 333°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Plastic bottle
Molecular Formula C8H8ClNO
Linear Formula CH3CONHC6H4Cl
Quantity 50 g
Beilstein 12, 611
Merck Index 14, 2111
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol,ether and carbon disulfide,slightly soluble in ccl4 and benzene
Molecular Weight (g/mol) 169.61
Formula Weight 169.61
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4′-Chloroacetanilide
Show More Show Less
compliance-icons
Product Identifier
  • 4'-Chloroacetanilide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.