missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-O-tolylhydrazine hydrochloride, 97%, Thermo Scientific™

Product Code. 10072404
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10072404 25 g 25g
10169222 1 g 1g
10022422 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10072404 Supplier Thermo Scientific Maybridge Supplier No. SB01504EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 19690-59-6
Molecular Formula C7H10Cl2N2
Molecular Weight (g/mol) 193.071
MDL Number MFCD00012944
InChI Key VBVZXWFQHFXJJK-UHFFFAOYSA-N
Synonym 4-chloro-2-methylphenyl hydrazine hydrochloride, 1-4-chloro-2-methylphenyl hydrazine hydrochloride, 4-chloro-2-tolylhydrazine hydrochloride, 4-chloro-2-methylphenylhydrazine, chloride, 4-chloro-2-tolylhydrazine hcl, 5-chloro-2-hydrazinotoluene hydrochloride, hydrazine, 4-chloro-2-methylphenyl-, hydrochloride 1:1
PubChem CID 3084321
IUPAC Name (4-chloro-2-methylphenyl)hydrazine;hydrochloride
SMILES CC1=C(C=CC(=C1)Cl)NN.Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Linear Formula ClC6H3(CH3)NHNH2·HCl
Quantity 25 g
Formula Weight 193.08
Percent Purity 97%
Chemical Name or Material 1-(4-Chloro-2-methylphenyl)hydrazine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.