missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-o-phenylenediamine, 97%

Product Code. 11459772
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459772 100 g 100g
11449772 25 g 25g
11469772 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11459772 Supplier Thermo Scientific Alfa Aesar Supplier No. A10823.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 95-83-0 | C6H7ClN2 | 142.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-83-0
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.59
MDL Number MFCD00011691
InChI Key BXIXXXYDDJVHDL-UHFFFAOYSA-N
Synonym 4-chloro-1,2-diaminobenzene, 4-chloro-o-phenylenediamine, 4-chloro-1,2-phenylenediamine, 1,2-diamino-4-chlorobenzene, 1,2-benzenediamine, 4-chloro, ursol olive 6g, 3,4-diaminochlorobenzene, 4-chloro-1,2-benzenediamine, 2-amino-4-chloroaniline, p-chloro-o-phenylenediamine
PubChem CID 7263
ChEBI CHEBI:82301
IUPAC Name 4-chlorobenzene-1,2-diamine
SMILES NC1=CC=C(Cl)C=C1N

Specifications

Melting Point 70°C to 74°C
Quantity 100 g
Formula Weight 142.59
Percent Purity 97%
Chemical Name or Material 4-Chloro-o-phenylenediamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.