missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-N-methylbenzylamine, 95%

Product Code. 15419928
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15419928 1 g 1g
15429928 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15419928 Supplier Alfa Aesar™ Supplier No. H63294.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 104-11-0
Molecular Formula C8H10ClN
Molecular Weight (g/mol) 155.625
MDL Number MFCD00018749
InChI Key LMBUJNXYGGNSAH-UHFFFAOYSA-N
Synonym 4-chloro-n-methylbenzylamine, n-methyl-4-chlorobenzylamine, 1-4-chlorophenyl-n-methylmethanamine, n-4-chlorobenzyl-n-methylamine, 4-chloro-benzyl-methyl-amine, 4-chlorophenyl methyl methyl amine, 4-chlorobenzyl methylamine, benzenemethanamine, 4-chloro-n-methyl, 4-chlorobenzylmethylamine, p-chloro-n-methylbenzylamine
PubChem CID 66905
IUPAC Name 1-(4-chlorophenyl)-N-methylmethanamine
SMILES CNCC1=CC=C(C=C1)Cl

Specifications

Boiling Point 77°C to 78°C (1 mmHg)
Quantity 1 g
UN Number UN2735
Formula Weight 155.62
Percent Purity 95%
Chemical Name or Material 4-Chloro-N-methylbenzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.