missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-N-(3-methylphenyl)butyramide, 97%, Thermo Scientific™

Product Code. 15414578
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15414578 500 mg 500mg
15404578 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15414578 Supplier Thermo Scientific Alfa Aesar Supplier No. H58249.ME
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73863-44-2
Molecular Formula C11H14ClNO
Molecular Weight (g/mol) 211.69
MDL Number MFCD00086969
InChI Key GBYXVHYDSWAERN-UHFFFAOYSA-N
Synonym 4-chloro-n-3-methylphenyl butanamide, butanamide, 4-chloro-n-3-methylphenyl, 4-chloro-n-m-tolyl butanamide, n-4-chlorobutyryl-m-toluidine, 4-chloro-meta-butyrotoluidide, 4-chloro-n-3-methylphenyl butyramide
PubChem CID 1809759
IUPAC Name 4-chloro-N-(3-methylphenyl)butanamide
SMILES CC1=CC=CC(NC(=O)CCCCl)=C1

Specifications

Quantity 500 mg
Formula Weight 211.69
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Chloro-N-(3-methylphenyl)butyramide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.