missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-N-(2-ethylhexyl)-3-nitrobenzamide, 97%, Thermo Scientific™

Catalog Number 15454826
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
449794-75-6
C15H21ClN2O3
312.79
MFCD01182374
XOGLBFUDOWUBEC-UHFFFAOYNA-N
4-chloro-n-2-ethylhexyl-3-nitrobenzamide
4095209
4-chloro-N-(2-ethylhexyl)-3-nitrobenzamide
CCCCC(CC)CNC(=O)C1=CC(=C(Cl)C=C1)[N+]([O-])=O
This item is not returnable. View return policy
449794-75-6
C15H21ClN2O3
312.79
MFCD01182374
XOGLBFUDOWUBEC-UHFFFAOYNA-N
4-chloro-n-2-ethylhexyl-3-nitrobenzamide
4095209
4-chloro-N-(2-ethylhexyl)-3-nitrobenzamide
CCCCC(CC)CNC(=O)C1=CC(=C(Cl)C=C1)[N+]([O-])=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 312.79
Percent Purity 97%
Chemical Name or Material 4-Chloro-N-(2-ethylhexyl)-3-nitrobenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'