missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-N-(2,6-dichlorophenyl)benzamide, 97%, Thermo Scientific™

Catalog Number 15455228
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
200807-46-1
C13H8Cl3NO
300.563
MFCD00028788
MGCCUXCCTBKJAN-UHFFFAOYSA-N
4-chloro-n-2,6-dichlorophenyl benzamide, 2',4,6'-trichlorobenzanilide
4693001
4-chloro-N-(2,6-dichlorophenyl)benzamide
C1=CC(=C(C(=C1)Cl)NC(=O)C2=CC=C(C=C2)Cl)Cl
This item is not returnable. View return policy
200807-46-1
C13H8Cl3NO
300.563
MFCD00028788
MGCCUXCCTBKJAN-UHFFFAOYSA-N
4-chloro-n-2,6-dichlorophenyl benzamide, 2',4,6'-trichlorobenzanilide
4693001
4-chloro-N-(2,6-dichlorophenyl)benzamide
C1=CC(=C(C(=C1)Cl)NC(=O)C2=CC=C(C=C2)Cl)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 300.57
Percent Purity 97%
Chemical Name or Material 4-Chloro-N-(2,6-dichlorophenyl)benzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'