missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-DL-phenylglycine, 98%

Product Code. 15476368
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15476368 25 g 25g
15486368 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15476368 Supplier Thermo Scientific Alfa Aesar Supplier No. H59921.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6212-33-5 | C8H8ClNO2 | 185.607 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in 0.1mol/L NaOH (almost transparency).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6212-33-5
Molecular Formula C8H8ClNO2
Molecular Weight (g/mol) 185.607
MDL Number MFCD00049328
InChI Key QGJGBYXRJVIYGA-UHFFFAOYSA-N
Synonym 2-amino-2-4-chlorophenyl acetic acid, amino 4-chlorophenyl acetic acid, amino-4-chloro-phenyl-acetic acid, dl-2-4-chlorophenyl glycine, 2-4-chlorophenyl glycine, +/--4-chlorophenylglycine, h-dl-phg 4-cl-oh, 4-chloro-dl-phenylglycine, dl-4-chlorophenyl glycine, 4-chlorophenyl glycine
PubChem CID 587821
IUPAC Name 2-amino-2-(4-chlorophenyl)acetic acid
SMILES C1=CC(=CC=C1C(C(=O)O)N)Cl

Specifications

Color White
Solubility Information Soluble in 0.1mol/L NaOH (almost transparency).
Quantity 25 g
Formula Weight 185.61
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Chloro-DL-phenylglycine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.