missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-DL-phenylalanine, 98+%
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Description
4-Chloro-DL-phenylalanine is an irreversible tryptophan hydroxylase inhibitor that depletes serotonin in the brain.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Chloro-DL-phenylalanine is an irreversible tryptophan hydroxylase inhibitor that depletes serotonin in the brain.
Solubility
Soluble to 10 mM in 1eq. NaOH and to 5 mM in water with gentle warming.
Notes
Store at room temperature. Incompatible with oxidizing agents.
4-Chloro-DL-phenylalanine is an irreversible tryptophan hydroxylase inhibitor that depletes serotonin in the brain.
Solubility
Soluble to 10 mM in 1eq. NaOH and to 5 mM in water with gentle warming.
Notes
Store at room temperature. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | ∼265°C (decomposition) |
| Beilstein | 2805758 |
| Solubility Information | Soluble to 10 mM in 1eq. NaOH and to 5 mM in water with gentle warming. |
| Quantity | 50 g |
| Formula Weight | 199.64 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 4-Chloro-DL-phenylalanine |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction