missing translation for 'onlineSavingsMsg'
Learn More

4-chloro-7-methylthieno[3,2-d]pyrimidine, 97%, Thermo Scientific™

Product Code. 10417184
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10417184 5 g 5g
10543032 1 g 1g
10131262 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10417184 Supplier Thermo Scientific Maybridge Supplier No. GK01634DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175137-21-0
Molecular Formula C7H5ClN2S
Molecular Weight (g/mol) 184.64
MDL Number MFCD00205202
InChI Key IYJDOVYAFDVIDB-UHFFFAOYSA-N
PubChem CID 2777586
IUPAC Name 4-chloro-7-methylthieno[3,2-d]pyrimidine
SMILES CC1=CSC2=C(Cl)N=CN=C12

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Synonym 4-chloro-7-methylthieno 3,2-d pyrimidine, thieno 3,2-d pyrimidine, 4-chloro-7-methyl, 4-chloro-7-methylthiopheno 3,2-d pyrimidine, thieno 3,2-d pyrimidine,4-chloro-7-methyl, pubchem23125, 4-chloro-7-methyl thieno 3,2-d pyrimidine, 4-chloro-7-methyl-thieno 3,2-d pyrimidine, 4-chloranyl-7-methyl-thieno 3,2-d pyrimidine, 4-chloro-7-methylthieno 2,3-d pyrimidine
Formula Weight 184.65
Percent Purity 97%
Chemical Name or Material 4-Chloro-7-methylthieno[3, 2-d]pyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.