missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-6-hydroxypyrimidine, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 4-Chloro-6-hydroxypyrimidine |
| CAS | 4765-77-9 |
| Molecular Formula | C4 H3 Cl N2 O |
| Quantity | 10 g |
| InChI Formula | InChI=1S/C4H3ClN2O/c5-3-1-4(8)7-2-6-3/h1-2H,(H,6,7,8) |
| SMILES | Oc1cc(Cl)ncn1 |
| IUPAC Name | 6-chloropyrimidin-4-ol |
| Molecular Weight (g/mol) | 130.53 |
| Formula Weight | 129.9934 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction