missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-6,7-dimethoxyquinazoline, 98%

Product Code. p-7030399
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11482030 5 g 5g
11492030 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11482030 Supplier Thermo Scientific Alfa Aesar Supplier No. H28405.06
Add to basket
This item is not returnable. View return policy

CAS: 13790-39-1 | C10H9ClN2O2 | 224.64 g/mol

A useful synthetic intermediate in the preparation of epidermal growth factors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A useful synthetic intermediate in the preparation of epidermal growth factors.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13790-39-1
Molecular Formula C10H9ClN2O2
Molecular Weight (g/mol) 224.64
MDL Number MFCD01570172
InChI Key LLLHRNQLGUOJHP-UHFFFAOYSA-N
Synonym 4-chloro-6,7-dimethoxy-quinazoline, quinazoline, 4-chloro-6,7-dimethoxy, 4-cholro-6,7-dimethoxyquinazoline, 6,7-dimethoxy-4-chloroquinazoline, zlchem 87, pubchem9672, acmc-1brd3, 4-chloro-6,7dimethoxyquinazoline, 4-chloro6,7-dimethoxyquinazoline, 4chloro-6,7-dimethoxyquinazoline
PubChem CID 2769364
IUPAC Name 4-chloro-6,7-dimethoxyquinazoline
SMILES COC1=C(OC)C=C2C(Cl)=NC=NC2=C1

Specifications

Melting Point 184°C to 187°C
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 224.65
Percent Purity 98%
Chemical Name or Material 4-Chloro-6,7-dimethoxyquinazoline
compliance-icons
Product Identifier
  • 4-Chloro-6,7-dimethoxyquinazoline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 4-Chloro-6,7-dimethoxyquinazoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.