missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-5-sulfamoyl-salicylic Acid, TRC

Product Code. 30003912
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003912 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30003912 Supplier Toronto Research Chemicals Supplier No. TRCC380025500MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14556-98-0
Molecular Formula C7 H6 Cl N O5 S
Molecular Weight (g/mol) 251.64
IUPAC Name 4-chloro-2-hydroxy-5-sulfamoylbenzoic acid
SMILES NS(=O)(=O)c1cc(C(=O)O)c(O)cc1Cl

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C7H6ClNO5S/c8-4-2-5(10)3(7(11)12)1-6(4)15(9,13)14/h1-2,10H,(H,11,12)(H2,9,13,14)
Formula Weight 250.9655
Chemical Name or Material 4-Chloro-5-sulfamoyl-salicylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.