missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-5-methylbenzene-1,2-diamine, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Synonym | 1,2-benzenediamine,4-chloro-5-methyl, 4-chloro-5-methyl-1,2-phenylenediamine, 4mrh, pubchem22761, 2-chloro-4,5-diaminotoluene, 2-amino-4-chloro-5-methylphenyl amine, 4-chloro-5-methyl-benzene-1,2-diamine, 5-chloro-4-methyl-1,2-phenylenediamine, 1,2-benzenediamine, 4-chloro-5-methyl, 4-chloranyl-5-methyl-benzene-1,2-diamine |
Formula Weight | 156.62 |
Percent Purity | 95% |
Quantity | 5 g |
Chemical Name or Material | 4-Chloro-5-methylbenzene-1, 2-diamine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'