Learn More
4-chloro-5-(2-thienyl)thieno[2,3-d]pyrimidine, 97%, Maybridge™
Brand: Maybridge AW00007DA
Code : 31
Additional Details : CAS Number : 189681-04-7 Weight : 0.05100kg
Quantity | 1g |
---|
Chemical Identifiers
189681-04-7 | |
252.734 | |
VNWBUFWVNCUUKY-UHFFFAOYSA-N | |
735732 | |
C1=CSC(=C1)C2=CSC3=C2C(=NC=N3)Cl |
C10H5ClN2S2 | |
MFCD00174019 | |
4-chloro-5-2-thienyl thieno 2,3-d pyrimidine, 4-chloro-5-thiophen-2-yl thieno 2,3-d pyrimidine, thieno 2,3-d pyrimidine,4-chloro-5-2-thienyl, 4-chloro-5-2-thienyl thiopheno 2,3-d pyrimidine, 4-chloro-5-thiophen-2-ylthieno 2,3-d pyrimidine | |
4-chloro-5-thiophen-2-ylthieno[2,3-d]pyrimidine |
Specifications
4-Chloro-5-(2-thienyl)thieno[2, 3-d]pyrimidine | |
1g | |
94.5 | |
95% | |
MFCD00174019 | |
VNWBUFWVNCUUKY-UHFFFAOYSA-N | |
4-chloro-5-thiophen-2-ylthieno[2,3-d]pyrimidine | |
735732 | |
95% |
Amber Glass Bottle | |
189681-04-7 | |
100.0 | |
C10H5ClN2S2 | |
4-chloro-5-2-thienyl thieno 2,3-d pyrimidine, 4-chloro-5-thiophen-2-yl thieno 2,3-d pyrimidine, thieno 2,3-d pyrimidine,4-chloro-5-2-thienyl, 4-chloro-5-2-thienyl thiopheno 2,3-d pyrimidine, 4-chloro-5-thiophen-2-ylthieno 2,3-d pyrimidine | |
C1=CSC(=C1)C2=CSC3=C2C(=NC=N3)Cl | |
252.734 | |
252.75 |