missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-4'-fluorobutyrophenone, 97%

Product Code. 10375690 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10375690 25 g Glass bottle 25g
10733831 100 g Glass bottle 100g
10122710 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10375690 Supplier Thermo Scientific Acros Supplier No. 109230250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3874-54-2 | C10H10ClFO | 200.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3874-54-2
Molecular Formula C10H10ClFO
Molecular Weight (g/mol) 200.64
MDL Number MFCD00001007
InChI Key HXAOUYGZEOZTJO-UHFFFAOYSA-N
Synonym 4-chloro-4'-fluorobutyrophenone, 4-chloro-1-4-fluorophenyl butan-1-one, 3-4-fluorobenzoyl propyl chloride, 4-fluorobenzoylpropyl chloride, 4-chloro-p-fluorobutyrophenone, 3-chloropropyl 4-fluorophenyl ketone, 1-butanone, 4-chloro-1-4-fluorophenyl, 4'-fluoro-4-chlorobutyrophenone, p-fluorobenzoylpropyl chloride, p-fluoro-4-chlorobutyrophenone
PubChem CID 19750
IUPAC Name 4-chloro-1-(4-fluorophenyl)butan-1-one
SMILES C1=CC(=CC=C1C(=O)CCCCl)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 5.0°C to 6.0°C
Color Green-Yellow
Density 1.2300g/mL
Boiling Point 130.0°C to 132.0°C (1.3 mbar)
Flash Point 152°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4CO(CH2)3Cl
Refractive Index 1.5245 to 1.5265
Quantity 25 g
Beilstein 07,IV,711
Specific Gravity 1.23
Solubility Information Solubility in water: 0.38g/L water (20°C). Other solubilities: soluble in methanol,ethanol,2-propanol,n,n-dmf,,acetone and dichloromethane
Formula Weight 200.64
Percent Purity 97%
Physical Form Oil
Chemical Name or Material 4-Chloro-4'-fluorobutyrophenone, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chloro-4'-fluorobutyrophenone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.