missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-(trifluoromethyl)pyridine hydrochloride, 97%, Thermo Scientific™

Product Code. 15214398 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15214398 5 g 5g
15204398 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15214398 Supplier Thermo Scientific Acros Supplier No. 454770050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 732306-24-0
Molecular Formula C6H4Cl2F3N
Molecular Weight (g/mol) 218
MDL Number MFCD04972770
InChI Key HCKXJUZITVCDND-UHFFFAOYSA-N
Synonym 4-chloro-3-trifluoromethyl pyridine hydrochloride, 4-chloro-3-trifluoromethyl pyridinehydrochloride, 4-chloro-3-trifluoromethyl pyridine, chloride, 4-chloro-3-trifluoromethyl pyridine hcl, 4-chloro-3-trifluoromethylpyridine hydrochloride, 3-chloro-4-trifluoromethyl pyridine hydrochlrorid, 4-chloranyl-3-trifluoromethyl pyridine hydrochloride, 4-chloro-3-trifluoromethylpyridine hydrochloride salt
PubChem CID 2782815
SMILES Cl.FC(F)(F)C1=C(Cl)C=CN=C1

Specifications

Melting Point 152.0°C to 154.0°C
Infrared Spectrum Authentic
Assay Percent Range 96.0 to max. 104% min. (Argentometry)
Quantity 5 g
IUPAC Name 4-chloro-3-(trifluoromethyl)pyridine hydrochloride
Formula Weight 218.01
Percent Purity 0.97
Chemical Name or Material 4-Chloro-3-(trifluoromethyl)pyridine hydrochloride
compliance-icons
Product Identifier
  • 4-Chloro-3-(trifluoromethyl)pyridine hydrochloride, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.