missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-(trifluoromethyl)phenyl isothiocyanate, 97%

Product Code. 11398235
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398235 1 g 1g
11308245 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398235 Supplier Thermo Scientific Alfa Aesar Supplier No. L10127.03
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 23163-86-2 | C8H3ClF3NS | 237.624 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23163-86-2
Molecular Formula C8H3ClF3NS
Molecular Weight (g/mol) 237.624
MDL Number MFCD00041090
InChI Key AHFPRSSHNSGRCU-UHFFFAOYSA-N
Synonym 4-chloro-3-trifluoromethyl phenyl isothiocyanate, 1-chloro-4-isothiocyanato-2-trifluoromethyl benzene, 4-chloro-3-trifluoromethyl phenylisothiocyanate, 4-chloro-3-trifluoromethylphenyl isothiocyanate, 1-chloro-4-isothiocyanato-2-trifluoromethyl-benzene, benzene, 1-chloro-4-isothiocyanato-2-trifluoromethyl, 4-chloro-3-trifluoromethyl benzenisothiocyanate, acmc-20amlf, pubchem5016, 4-chloro-3-trifluoromethylphenylisothiocyanate
PubChem CID 520026
IUPAC Name 1-chloro-4-isothiocyanato-2-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1N=C=S)C(F)(F)F)Cl

Specifications

Density 1.47
Boiling Point 248°C to 251°C
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.59
Quantity 1 g
UN Number UN2922
Beilstein 787371
Sensitivity Moisture sensitive
Formula Weight 237.63
Percent Purity 97%
Chemical Name or Material 4-Chloro-3-(trifluoromethyl)phenyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.