missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-(trifluoromethyl)phenyl isocyanate, 98%

Product Code. 15720859 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15720859 25 g Glass bottle 25g
10503231 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15720859 Supplier Thermo Scientific Acros Supplier No. 309130250
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 327-78-6 | C8H3ClF3NO | 221.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 327-78-6
Molecular Formula C8H3ClF3NO
Molecular Weight (g/mol) 221.56
MDL Number MFCD00013874
InChI Key NBJZEUQTGLSUOB-UHFFFAOYSA-N
Synonym 4-chloro-3-trifluoromethyl phenyl isocyanate, 1-chloro-4-isocyanato-2-trifluoromethyl benzene, 4-chloro-3-trifluoromethyl phenylisocyanate, benzene, 1-chloro-4-isocyanato-2-trifluoromethyl, 4-chloro-3-trifluoromethylphenyl isocyanate, 4-chloro-3-trifluoromethoxy phenyl isocyanate, 1-chloro-4-isocyanato-2-trifluoromethyl-benzene, 4-chloro-3-trifluoromethyl benzenisocyanate, 3-trifluoromethyl-4-chloro-phenyl isocyanate, pubchem5027
PubChem CID 2733265
IUPAC Name 1-chloro-4-isocyanato-2-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1N=C=O)C(F)(F)F)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 29.0°C to 37.0°C
Color White to Yellow
Flash Point 99°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H3(CF3)NCO
Quantity 25 g
Formula Weight 221.56
Percent Purity 98%
Physical Form Low Melting Solid, Powder or Chunks
Chemical Name or Material 4-Chloro-3-(trifluoromethyl)phenyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.