missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-(trifluoromethyl)phenyl isocyanate, 97%

Product Code. 11338565
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338565 5 g 5g
11328565 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11338565 Supplier Thermo Scientific Alfa Aesar Supplier No. L12016.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 327-78-6 | C8H3ClF3NO | 221.563 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 327-78-6
Molecular Formula C8H3ClF3NO
Molecular Weight (g/mol) 221.563
MDL Number MFCD00013874
InChI Key NBJZEUQTGLSUOB-UHFFFAOYSA-N
Synonym 4-chloro-3-trifluoromethyl phenyl isocyanate, 1-chloro-4-isocyanato-2-trifluoromethyl benzene, 4-chloro-3-trifluoromethyl phenylisocyanate, benzene, 1-chloro-4-isocyanato-2-trifluoromethyl, 4-chloro-3-trifluoromethylphenyl isocyanate, 4-chloro-3-trifluoromethoxy phenyl isocyanate, 1-chloro-4-isocyanato-2-trifluoromethyl-benzene, 4-chloro-3-trifluoromethyl benzenisocyanate, 3-trifluoromethyl-4-chloro-phenyl isocyanate, pubchem5027
PubChem CID 2733265
IUPAC Name 1-chloro-4-isocyanato-2-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1N=C=O)C(F)(F)F)Cl

Specifications

Melting Point 34°C to 36°C
Boiling Point 86°C to 90°C (14 mmHg)
Flash Point 99°C (210°F)
Odor Pungent
Quantity 5 g
UN Number UN2811
Beilstein 522032
Sensitivity Moisture sensitive
Formula Weight 221.57
Percent Purity 97%
Chemical Name or Material 4-Chloro-3-(trifluoromethyl)phenyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.