missing translation for 'onlineSavingsMsg'
Learn More

4-chloro-3-(trifluoromethyl)benzaldehyde, 98%, Thermo Scientific™

Product Code. 10524954 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10524954

Brand: Thermo Scientific Acros 425840010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 34328-46-6
CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.4500g/mL
Boiling Point 218°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C8H4ClF3O
Linear Formula ClC6H3(CF3)CHO
MDL Number MFCD00040722
Quantity 1 g
Specific Gravity 1.45
Synonym 4-chloro-3-trifluoromethyl benzaldehyde, 4-chloro-3-trifluoromethyl-benzaldehyde, 2-chloro-5-formylbenzotrifluoride, benzaldehyde, 4-chloro-3-trifluoromethyl, pubchem2892, acmc-1adw8, timtec-bb sbb003468, 4-chloro-3trifluoromethyl-benzaldehyde, 3-trifluoromethyl-4-chloro-benzaldehyde, 4-chloranyl-3-trifluoromethyl benzaldehyde
InChI Key NIHMMULLFBKTOK-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1C=O)C(F)(F)F)Cl
IUPAC Name 4-chloro-3-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol) 208.57
PubChem CID 118642
Formula Weight 208.57
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-chloro-3-(trifluoromethyl)benzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chloro-3-(trifluoromethyl)benzaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.