missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-(trifluoromethyl)aniline, 99%

Product Code. 11369709
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11369709 100 g 100g
11349709 5 g 5g
11359709 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11369709 Supplier Thermo Scientific Alfa Aesar Supplier No. B21200.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 320-51-4 | C7H5ClF3N | 195.569 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 320-51-4
Molecular Formula C7H5ClF3N
Molecular Weight (g/mol) 195.569
MDL Number MFCD00007846
InChI Key ASPDJZINBYYZRU-UHFFFAOYSA-N
Synonym 4-chloro-3-trifluoromethyl aniline, 5-amino-2-chlorobenzotrifluoride, 3-trifluoromethyl-4-chloroaniline, 4-chloro-alpha,alpha,alpha-trifluoro-m-toluidine, 2-chloro-5-aminobenzotrifluoride, acbt, benzenamine, 4-chloro-3-trifluoromethyl, 2-chloro-5-aminebenzotrifluoride, 4-chloro-3-trifluoromethyl-phenylamine, 4-chloro-3-trifluoromethyl benzenamine
PubChem CID 67574
ChEBI CHEBI:59245
IUPAC Name 4-chloro-3-(trifluoromethyl)aniline
SMILES C1=CC(=C(C=C1N)C(F)(F)F)Cl

Specifications

Melting Point 35°C to 38°C
Boiling Point 85°C (3 mmHg)
Flash Point 134°C (273°F)
Quantity 100 g
UN Number UN2811
Beilstein 641588
Formula Weight 195.57
Percent Purity 99%
Chemical Name or Material 4-Chloro-3-(trifluoromethyl)aniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.