missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-sulfamoylbenzoic Acid, Mikromol™

Product Code. 17951406
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17951406 25 mg 25mg
17921646 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17951406 Supplier Mikromol Supplier No. MM0477.040025
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-Chloro-3-sulfamoylbenzoic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Chloro-3-sulfamoylbenzoic Acid
CAS 1205-30-7
Analyte or Component Names 4-Chloro-3-sulfamoylbenzoic Acid
Shipping Condition Room Temperature
Molecular Formula C7 H6 Cl N O4 S
Certifications/Compliance ISO 17025
Synonym 4-Chloro-3-sulfamoylbenzoic Acid, Indapamide Related Compound E, Clopamide Impurity C
InChI Formula InChI=1S/C7H6ClNO4S/c8-5-2-1-4(7(10)11)3-6(5)14(9,12)13/h1-3H,(H,10,11)(H2,9,12,13)
SMILES NS(=O)(=O)c1cc(ccc1Cl)C(=O)O
IUPAC Name 4-chloro-3-sulfamoylbenzoic acid
Molecular Weight (g/mol) 235.64
Quantity 25 mg
Formula Weight 234.9706 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.