missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-sulfamoylbenzoic acid, 98%

Product Code. 11461037
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11461037 50 g 50g
11471037 250 g 250g
11451037 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11461037 Supplier Thermo Scientific Alfa Aesar Supplier No. B24057.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1205-30-7 | C7H6ClNO4S | 235.638 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1205-30-7
Molecular Formula C7H6ClNO4S
Molecular Weight (g/mol) 235.638
MDL Number MFCD00012375
InChI Key FHQAWINGVCDTTG-UHFFFAOYSA-N
Synonym 4-chloro-5-sulphamoylbenzoic acid, 3-aminosulfonyl-4-chlorobenzoic acid, csba, sulfamido-3-chlorobenzoic acid, benzoic acid, 3-aminosulfonyl-4-chloro, 3-sulfamoyl-4-chlorobenzoic acid, benzoic acid, 4-chloro-3-sulfamoyl, 4-chloro-3-sulfamoyl-benzoic acid, 4-chloro-5-sulfamoylbenzoic acid, 4-chloro-3-sulphamoylbenzoic acid
PubChem CID 14568
IUPAC Name 4-chloro-3-sulfamoylbenzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)S(=O)(=O)N)Cl

Specifications

Melting Point 255°C to 260°C
Quantity 50 g
Formula Weight 235.65
Percent Purity 98%
Chemical Name or Material 4-Chloro-3-sulfamoylbenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.