missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-nitrotoluene, 97+%

Product Code. 11400213
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400213 2500 g 2500g
11480203 100 g 100g
11490203 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11400213 Supplier Thermo Scientific Alfa Aesar Supplier No. A11383.0E
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 89-60-1 | C7H6ClNO2 | 171.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-60-1
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007085
InChI Key NWESJZZPAJGHRZ-UHFFFAOYSA-N
Synonym 4-chloro-3-nitrotoluene, 3-nitro-4-chlorotoluene, benzene, 1-chloro-4-methyl-2-nitro, 2-chloro-5-methylnitrobenzene, 1-chloro-4-methyl-2-nitro-benzene, toluene, 4-chloro-3-nitro, 3-nitro-4-chlorotoluol, pubchem19752, 4,3-chloronitrotoluene, 4-chloro-3-nitro-toluene
PubChem CID 6976
IUPAC Name 1-chloro-4-methyl-2-nitrobenzene
SMILES CC1=CC=C(Cl)C(=C1)[N+]([O-])=O

Specifications

Melting Point ∼7°C
Density 1.295
Boiling Point 260°C to 262°C
Flash Point 129°C (264°F)
Refractive Index 1.558
Quantity 2500 g
UN Number UN2433
Beilstein 511055
Formula Weight 171.58
Percent Purity ≥97%
Chemical Name or Material 4-Chloro-3-nitrotoluene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.