missing translation for 'onlineSavingsMsg'
Learn More

4-chloro-3-nitrophenyl isothiocyanate, Thermo Scientific™

Product Code. 10016662
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10016662 10 g 10g
10506035 5 g 5g
10162884 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10016662 Supplier Thermo Scientific Maybridge Supplier No. SB01784EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 127142-66-9
Molecular Formula C7H3ClN2O2S
Molecular Weight (g/mol) 214.623
InChI Key ZXGZBHIDSJXKLE-UHFFFAOYSA-N
Synonym 4-chloro-3-nitrophenyl isothiocyanate, 4-chloro-3-nitrophenylisothiocyanate, acmc-1c0qb, 4-chloro-3-nitrobenzenisothiocyanate, #, benzene,1-chloro-4-isothiocyanato-2-nitro, 1-chloranyl-4-isothiocyanato-2-nitro-benzene
PubChem CID 145581
IUPAC Name 1-chloro-4-isothiocyanato-2-nitrobenzene
SMILES C1=CC(=C(C=C1N=C=S)[N+](=O)[O-])Cl

Specifications

Assay Percent Range 97%
Quantity 10 g
Formula Weight 214.63
Percent Purity 97%
Chemical Name or Material 4-Chloro-3-nitrophenyl isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.