missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-3-nitrophenol, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 4-Chloro-3-nitrophenol |
| CAS | 610-78-6 |
| Molecular Formula | C6H4ClNO3 |
| Quantity | 1 g |
| Synonym | 2-Chloro-5-hydroxynitrobenzene, 3-Nitro-4-chlorophenol, NSC 211001 |
| InChI Formula | InChI=1S/C6H4ClNO3/c7-5-2-1-4(9)3-6(5)8(10)11/h1-3,9H |
| SMILES | Oc1ccc(Cl)c(c1)[N+](=O)[O-] |
| IUPAC Name | 4-chloro-3-nitrophenol |
| Molecular Weight (g/mol) | 173.55 |
| Formula Weight | 172.988 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction