missing translation for 'onlineSavingsMsg'
Learn More

4-chloro-3-nitrophenol, 98%, Thermo Scientific™

Product Code. 10611653 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
25g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10611653 25g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10611653 Supplier Thermo Scientific Acros Supplier No. 369590250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 610-78-6
CAS Min % 97.5
CAS Max % 100.0
Melting Point 125°C to 129°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C6H4ClNO3
Linear Formula ClC6H3(NO2)OH
MDL Number MFCD00043546
Quantity 25 g
Beilstein 06, IV, 1352
Synonym phenol, 4-chloro-3-nitro, 4-chloro-3-nitro-phenol, 4chloro-3-nitrophenol, pubchem10459, 4-chlor-3-nitro-phenol, 3-nitro-4-chloro phenol, acmc-1azj5, phenol,4-chloro-3-nitro, 4-chloranyl-3-nitro-phenol, 2-chloro-5-hydroxy nitrobenzene
InChI Key JUIKCULGDIZNDI-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1O)[N+](=O)[O-])Cl
IUPAC Name 4-chloro-3-nitrophenol
Molecular Weight (g/mol) 173.56
PubChem CID 69127
Formula Weight 173.56
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-chloro-3-nitrophenol
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chloro-3-nitrophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.