missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-nitrobenzotrifluoride, TRC

Product Code. 30180188
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30180188 25 g 25g
30179774 5 g 5g
30180167 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180188 Supplier Toronto Research Chemicals Supplier No. TRCC38059025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-17-5
Molecular Formula C7 H3 Cl F3 N O2
Molecular Weight (g/mol) 225.55
Synonym 4-Chloro-3-nitrobenzotrifluoride, 4-Chloro-α,α,α-trifluoro-3-nitro-toluene, (3-Nitro-4-chlorophenyl)trifluoromethane;
IUPAC Name 1-chloro-2-nitro-4-(trifluoromethyl)benzene
SMILES [O-][N+](=O)c1cc(ccc1Cl)C(F)(F)F

Specifications

Quantity 25 g
InChI Formula InChI=1S/C7H3ClF3NO2/c8-5-2-1-4(7(9,10)11)3-6(5)12(13)14/h1-3H
Formula Weight 224.9804
Chemical Name or Material 4-Chloro-2-nitrobenzotrifluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.