missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-nitrobenzaldehyde, 97%

Product Code. 11470034
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11470034 10 g 10g
11480034 50 g 50g
11490034 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11470034 Supplier Thermo Scientific Alfa Aesar Supplier No. A13549.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16588-34-4 | C7H4ClNO3 | 185.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16588-34-4
Molecular Formula C7H4ClNO3
Molecular Weight (g/mol) 185.56
MDL Number MFCD00007078
InChI Key HETBKLHJEWXWBM-UHFFFAOYSA-N
Synonym 3-nitro-4-chlorobenzaldehyde, benzaldehyde, 4-chloro-3-nitro, chembl51664, 4-chloro-3-nitro-benzaldehyde, pubchem14274, acmc-1c7bn, 3nitro-4-chlorobenzaldehyde, asischem r36965, dsstox_cid_24846, dsstox_rid_80526
PubChem CID 85505
IUPAC Name 4-chloro-3-nitrobenzaldehyde
SMILES [O-][N+](=O)C1=CC(C=O)=CC=C1Cl

Specifications

Melting Point 61°C to 65°C
Quantity 10 g
Beilstein 778323
Sensitivity Air Sensitive
Formula Weight 185.57
Percent Purity 97%
Chemical Name or Material 4-Chloro-3-nitrobenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.