missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-nitro-2-pyridone, 98%

Product Code. 11352737
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11352737 5 g 5g
11362737 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352737 Supplier Thermo Scientific Alfa Aesar Supplier No. L15686.06
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 165547-79-5 | C5H3ClN2O3 | 174.54 g/mol

4-Chloro-3-nitro-2-pyridone finds an important application as raw material in organic synthesis and pharmaceuticals. It is also used as an intermediate in agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloro-3-nitro-2-pyridone finds an important application as raw material in organic synthesis and pharmaceuticals. It is also used as an intermediate in agrochemicals and dyestuffs.

Solubility
Insoluble in water.

Notes
Keep the container tightly sealed. Store in a cool, dry condition. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 165547-79-5
Molecular Formula C5H3ClN2O3
Molecular Weight (g/mol) 174.54
MDL Number MFCD03093067
InChI Key UKIZCTHOMJXNIX-UHFFFAOYSA-N
Synonym 4-chloro-2-hydroxy-3-nitropyridine, 4-chloro-3-nitropyridin-2-ol, 4-chloro-3-nitropyridone, 4-chloro-3-nitro-2-pyridone, 4-chloro-3-nitropyridin-2 1h-one, 4-chloro-3-nitro-pyridin-2-ol, 4-chloro-3-nitro-pyridin-2-one, 2 1h-pyridinone, 4-chloro-3-nitro, pubchem6547, acmc-209dsr
PubChem CID 519282
IUPAC Name 4-chloro-3-nitro-1H-pyridin-2-one
SMILES C1=CNC(=O)C(=C1Cl)[N+](=O)[O-]

Specifications

Melting Point 222°C to 224°C
Quantity 5 g
Beilstein 149557
Solubility Information Insoluble in water.
Formula Weight 174.54
Percent Purity 98%
Chemical Name or Material 4-Chloro-3-nitro-2-pyridone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.