missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-3-methylphenylmagnesium bromide, 0.5M in 2-MeTHF

Product Code. 15410706
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15410706 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15410706 Supplier Alfa Aesar™ Supplier No. H54052.AE

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 460747-53-9
Quantity 100 mL
Molecular Formula C7H6BrClMg
MDL Number MFCD02260216
UN Number UN2924
Synonym 4-chloro-3-methylphenylmagnesium bromide, 4-chloro-3-methylphenylmagnesium bromide, 0.5m in 2-methf, bromo 4-chloro-3-methylphenyl magnesium, prfxdtbctcbdkh-uhfffaoysa-m, magnesium,1-chloro-2-methylbenzene-4-ide,bromide, 4-chloro-3-methylphenylmagnesium bromide 0.5 m in tetrahydrofuran
InChI Key GRISSYATSPDQCA-UHFFFAOYSA-M
SMILES CC1=C[C-]=CC=C1Cl.[Mg+2].[Br-]
IUPAC Name magnesium;1-chloro-2-methylbenzene-4-ide;bromide
Molecular Weight (g/mol) 229.784
PubChem CID 24722687
Formula Weight 229.79
Concentration or Composition (by Analyte or Components) 0.5M in 2-MeTHF
Chemical Name or Material 4-Chloro-3-methylphenylmagnesium bromide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.