missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-methylphenol, TRC

Product Code. 30136285
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136285 250 g 250g
30135265 100 g 100g
17993895 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136285 Supplier Toronto Research Chemicals Supplier No. TRCC369765250G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-50-7
Molecular Formula C7 H7 Cl O
Molecular Weight (g/mol) 142.58
Synonym m-Cresol, 4-chloro- (8CI), 4-Chloro-3-methylphenol, 1-Chloro-2-methyl-4-hydroxybenzene, 2-Chloro-5-hydroxytoluene, 3-Methyl-4-chlorophenol, 3-Methyl-p-chlorophenol, 4-Chloro-3-cresol, 4-Chloro-5-methylphenol, 4-Chloro-m-cresol, 6-Chloro-3-hydroxytoluene, Aldecoc XD, Aptal, Baktol, Baktolan, Candaseptic, Chlorocresol, NSC 4166, Neopredisan, Ottafact, PCMC, Parmetol, Parol, Peritonan, Preventol CMK, Raschit, Raschit K, Rasen-Anicon, p-Chloro-m-cresol, para-Chloro-meta-cresol
IUPAC Name 4-chloro-3-methylphenol
SMILES Cc1cc(O)ccc1Cl

Specifications

Quantity 250 g
InChI Formula InChI=1S/C7H7ClO/c1-5-4-6(9)2-3-7(5)8/h2-4,9H,1H3
Formula Weight 142.0185
Percent Purity >95%
Chemical Name or Material 4-Chloro-3-methylphenol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.