missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-methylaniline, TRC

Product Code. 30138004
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138004 50 g 50g
30138319 25 g 25g
30008359 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138004 Supplier Toronto Research Chemicals Supplier No. TRCC36907550G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7149-75-9
Molecular Formula C7 H8 Cl N
Molecular Weight (g/mol) 141.6
IUPAC Name 4-chloro-3-methylaniline
SMILES Cc1cc(N)ccc1Cl

Specifications

Quantity 50 g
InChI Formula InChI=1S/C7H8ClN/c1-5-4-6(9)2-3-7(5)8/h2-4H,9H2,1H3
Formula Weight 141.0345
Chemical Name or Material 4-Chloro-3-methylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.