missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloro-3'-methylacetophenone, tech. 75%, remainder 3'-chloro-4'-methylacetophenone

Product Code. 11362326
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11362326 25 g 25g
11352326 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362326 Supplier Thermo Scientific Alfa Aesar Supplier No. L06011.14
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 37074-39-8 | C9H9ClO | 168.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37074-39-8
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD00045172
InChI Key XOIGZLJCLDWTQH-UHFFFAOYSA-N
Synonym 1-4-chloro-3-methylphenyl ethanone, 4'-chloro-3'-methylacetophenone, 4-chloro-3-methylacetophenone, ethanone, 1-4-chloro-3-methylphenyl, 1-4-chloro-3-methylphenyl ethan-1-one, 4-chloro-3-methyl acetophenone, 1-4-chloro-3-methyl-phenyl-ethanone, ksc577m5h, 3-methyl-4-chloroacetophenone, 2-methyl-4-acetyl-chlorbenzol
PubChem CID 592743
IUPAC Name 1-(4-chloro-3-methylphenyl)ethanone
SMILES CC(=O)C1=CC(C)=C(Cl)C=C1

Specifications

Density 1.15
Boiling Point 104°C to 105°C (0.4 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 774639
Formula Weight 168.62
Percent Purity 75%
Grade Technical
Assay remainder 3'-chloro-4'-methylacetophenone
Chemical Name or Material 4'-Chloro-3'-methylacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.