missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-fluoropyridine, 95%

Product Code. 11976451 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11976451 5 g 5g
11986451 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11976451 Supplier Thermo Scientific Acros Supplier No. 443160050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2546-56-7 | C5H3ClFN | 131.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2546-56-7
Molecular Formula C5H3ClFN
Molecular Weight (g/mol) 131.53
MDL Number MFCD03453233
InChI Key BEQUUSCRAKEKQM-UHFFFAOYSA-N
Synonym pyridine, 4-chloro-3-fluoro, 3-fluoro-4-chloro pyridine, 3-fluoro-4-chloropyridine hydrochloride, pubchem6667, 4-chloro-3fluoropyridine, 3-fluoro-4-chloropyridine, acmc-209x6p, 4-chloro-3-fluoro-pyridin, 3-fluoro-4-chloro-pyridine, 4-chloro-3-fluoro-pyridine
PubChem CID 3852429
IUPAC Name 4-chloro-3-fluoropyridine
SMILES FC1=C(Cl)C=CN=C1

Specifications

Melting Point -27°C to 24°C
Boiling Point 142°C to 143°C
Flash Point 28°C
Assay Percent Range 94% min. (GC)
Quantity 5 g
Formula Weight 131.54
Percent Purity 95%
Chemical Name or Material 4-Chloro-3-fluoropyridine
compliance-icons compliance-icons
Product Identifier
  • 4-Chloro-3-fluoropyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.